Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC CASTUS LINER CARTRDG 270 SHORT
Supplier:  ATLANTIC BIOPHARM SOLUTIONS LLC 18000002
Encompass_Preferred
Castus liner cartridge 270 short film: PE 80µm x 25m Ø 350 mmPacking in additional bag NS
Catalog No. NC2871936