Learn More
Aviva Systems Biology Corp CASZ1 Antibody - middle region (ARP33139_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33139_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CASZ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CASZ1. Alternate names: FLJ12223, FLJ20321, SRG, ZNF693, cst, dJ734G22.1, CST, RP4-734G22.2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54895