Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ CAT 2 Arc Flash Accessory Kit with Class 0 Gloves
SDP

Catalog No. 170011209 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1209 Small
17-001-1210 Medium
17-001-1211 Large
17-001-1212 X-Large
17-001-1213 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-001-1209 Supplier National Safety Apparel™ Supplier No. KIT2NC08PVB
Add to Cart
Add to Cart

With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability

  • Flat locked seams for comfort
  • Rib knit, single layer hood balaclava
  • Adjustable settings allow the faceshield to be secured in various positions
  • Lightweight design
  • PureView™ technology for proven color recognition

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Material Westex™ UltraSoft™
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, ASTM F2178, ANSI/ASTM D120, Class 0, Type 1, OSHA 1910.269
Product Type Accessory Kit
Gender Unisex
Arc Flash Rating 12 cal/cm²
Color Navy
Suspender None
Closure Type None
Size Small
Seam Type Stitched
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.