Learn More
World Precision Instrument Cat Adaptor for 505211
SDP
Catalog No. 502539171
Encompass
The Dog/Monkey Adaptor is available only for the Large Animal Stereotaxic, 4 Manipulator Arms (505326). To fit any size dog, it should be used with 505341 and 505342 stereotaxic instruments, which have two sets of ear bar holders at different spacings from the front, to accommodate a wide size range of dogs.
Catalog No. 50-253-9171 Supplier World Precision Instrument Supplier No. 505244
The Dog/Monkey Adaptor is available only for the Large Animal Stereotaxic, 4 Manipulator Arms (505326). To fit any size dog, it should be used with 505341 and 505342 stereotaxic instruments, which have two sets of ear bar holders at different spacings from the front, to accommodate a wide size range of dogs.