Learn More
World Precision Instrument Cat Adaptor, Non Rupture
SDP
Catalog No. 502539170
Encompass
The Dog/Monkey Adaptor is available only for the Large Animal Stereotaxic, 4 Manipulator Arms (505326). To fit any size dog, it should be used with 505341 and 505342 stereotaxic instruments, which have two sets of ear bar holders at different spacings from the front, to accommodate a wide size range of dogs.
Catalog No. 50-253-9170 Supplier World Precision Instrument Supplier No. 505245
The Dog/Monkey Adaptor is available only for the Large Animal Stereotaxic, 4 Manipulator Arms (505326). To fit any size dog, it should be used with 505341 and 505342 stereotaxic instruments, which have two sets of ear bar holders at different spacings from the front, to accommodate a wide size range of dogs.