Learn More
Sino Biological CAT / Catalase Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12084T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CAT / Catalase (rh CAT / Catalase; Catalog#12084-H07B; P04040; Ala2-Leu527). CAT / Catalase specific IgG was purified by Human CAT / Catalase affinity chromatography.
Catalog No. 50-224-8469