Learn More
World Precision Instrument Cat Ear Bars, 20 degree
SDP
Supplier:  World Precision Instrument 505332
Encompass
Ear Bars have laser-engraved scales, 50mm of calibration and are 6.2 mm square. WPI Mouse and Rat Stereotaxic Instruments and WPI Rat Stereotaxic Instruments Ear Bars have a different zero point than those for the 505326 series. Standard (18°) Ear Bars taper at the tip to penetrate far into the ear canal for a more secure hold. Non-puncture Ear Bars have a 45° taper and do not puncture the tympanic membrane.
Catalog No. 50-253-9114