Learn More
Biocare Medical, LLC CAT Hematoxylin, for Use in the Histologic Demonstration of Nuclear Staining, 500 ml
SDP
Catalog No. 5082393
Encompass_Preferred
CAT Hematoxylin is a modified Lillie-Mayer's Method formulation that provides incredible nuclear detail in routine H&E's and IHC, ard some special stains. It is formulated to eliminate the necessity for section differentiation.
Catalog No. 50-823-93 Supplier Biocare Medical, LLC Supplier No. CATHEM
CAT Hematoxylin is a modified Lillie-Mayer's Method formulation that provides incredible nuclear detail in routine H&E's and IHC, ard some special stains. It is formulated to eliminate the necessity for section differentiation.