Learn More
Immunology Consultants Cat IgG Purified, 5.0 mg
SDP
Supplier:  Immunology Consultants AG20G
Encompass_Preferred
The IgG h+l was purified from normal cat serum using a combination of procedures that include salt fractionation. Antibody concentration was determined by extinction coefficient: O.D. 1.4 at 280nM equals 1.0 mg of IgG. Estimated purity of >95% as determined by SDS-PAGE. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2161