Learn More
Immunology Consultants Cat IgM ELISA Kit
SDP
Supplier:  Immunology Consultants E20M
Encompass_Preferred
The Feline/Cat IgM test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgM in cat biological samples.
Catalog No. 50-240-6572