Learn More
Mabtech Inc CAT INTERFERON GAMMA ELISPOT K
Supplier:  Mabtech Inc 31222A
Encompass
Cat interferon gamma Elispot kit - basic CONTENTS: Capture mAb (MT131)Biotinylated detection mAb (MT114)Streptavidin-ALP;SIZE: for 4 plates
Catalog No. NC1517691