Learn More
Med Vet International Cat Nail Clipper, Stainless Steel, 3in, J0084C, Each
Catalog No. 5000584935
Encompass_Preferred
Cat Claw Scissors especially designed for cutting the thin and fragile nails of cats and small exotics. Gives a good clean cut without tearing or shredding.
Catalog No. 50-005-84935 Supplier Med Vet International Supplier No. JOR84C
Cat Claw Scissors especially designed for cutting the thin and fragile nails of cats and small exotics. Gives a good clean cut without tearing or shredding.