Learn More
Stoelting™ Cat/Rabbit Ventilator
SDP
Catalog No. 10001104 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Synchronized start/stop ventilator
Ventilator
Size:
50 mL
100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size
10-001-104 Ventilator 100 mL
10-001-072 Ventilator 50 mL
10-001-094 Synchronized start/stop ventilator 50 mL
10-000-954 Synchronized start/stop ventilator 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-001-104 Supplier Stoelting™ Supplier No. 5019510
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Cat/Rabbit Ventilator features a unique design that ensures accuracy, durability and quietness of operation.
  • Valve mechanism features nearly zero dead space and superb durability with no leakage. Expired air may be channeled for analysis and/or recycled, when used with anesthetic gases.
  • Digital stroke rate LED display
  • User selected pause (Model 56125) - can be paused at any point in the respiration cycle for taking measurements or for gas analysis.
  • Easily adjustable stroke volume whether the ventilator is on or off
    • For the most convenient and accurate stroke volume control available, the knob and scale remain stationary, even when the piston is in motion. 
  • Elimination of mechanical noise, reduced wear, and very low maintenance requirements
  • Cylinder/piston assemblies available
    • Cat units are supplied with one of two easily interchangeable cylinder/piston assemblies that enable a broader range of stroke volumes
Specifications
For Use With (Application) Neuroscience
Size 100 mL
Animal Cat, Rabbit
Model Ventilator