Learn More
Hach Company Cationic Surfactants TNTplus Vial Test (0.2 - 2.0 mg/L), 25 Tests
Catalog No. 501998166 Shop All Hach Company Products
Encompass
Cationic Surfactants TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime. For determination of lower concentrations of Cationic Surfactants by the Bromophenol Blue method, calibrated with CTAB. Traditional surfactants measurement has often involved time consuming handling steps, requiring expensive high end instrumentation or the use of large amounts of toxic chemistries. With Surfactants TNTplus reagents, simply add sample and included reagents to the vial, shake the vial for a defined period of time and insert into your Hach spectrophotometer! The Direct Read Technology will provide your Surfactants results in mg/L! No more cleaning glassware, dealing with large amounts of toxic waste, and worrying about human errors. 25 Cationic Surfactants vials per box.
Catalog No. 50-199-8166 Supplier Hach Company Supplier No. TNT885
Cationic Surfactants TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime. For determination of lower concentrations of Cationic Surfactants by the Bromophenol Blue method, calibrated with CTAB. Traditional surfactants measurement has often involved time consuming handling steps, requiring expensive high end instrumentation or the use of large amounts of toxic chemistries. With Surfactants TNTplus reagents, simply add sample and included reagents to the vial, shake the vial for a defined period of time and insert into your Hach spectrophotometer! The Direct Read Technology will provide your Surfactants results in mg/L! No more cleaning glassware, dealing with large amounts of toxic waste, and worrying about human errors. 25 Cationic Surfactants vials per box.