Learn More
Grainger CAT6 RJ45 Inline Coupler White
Catalog No. 502752713
Encompass_Preferred
The Monoprice Inline Coupler 7286 is a BNC inline coupler that allows you to connect two BNC cables together. It is nickel plated and has a 50 ohm impedance. The coupler has a frequency range up to 4GHz. It can handle up to 1kW of power. The dimensions are 0.47 x 0.47 x 1.26 inches. This is a high quality BNC coupler made by Monoprice that is useful for connecting BNC cables in video, CCTV, and other applications.
Catalog No. 50-275-2713 Supplier Grainger Supplier No. 14J358
The Monoprice Inline Coupler 7286 is a BNC inline coupler that allows you to connect two BNC cables together. It is nickel plated and has a 50 ohm impedance. The coupler has a frequency range up to 4GHz. It can handle up to 1kW of power. The dimensions are 0.47 x 0.47 x 1.26 inches. This is a high quality BNC coupler made by Monoprice that is useful for connecting BNC cables in video, CCTV, and other applications.