Learn More
Catachem Plasma Free Hemoglobin Control Level I, 3 x 3 mL, Liquid
SDP
Catalog No. NC0998352
Encompass
The CATACHEM HemoglobinCNTRL 1 is designed for monitoring the quality performance of plasma free hemoglobin determination procedures. This control material is essential for ensuring the accuracy and reliability of assays that measure free hemoglobin in plasma, particularly for diagnosing conditions like hemolytic transfusion reactions and intravascular hemolysis. Its ready-to-use liquid format simplifies laboratory workflows, eliminating the need for preparation. Laboratories are advised to use this control to maintain consistent quality performance in their analytical procedures.
  • Provides a known Plasma Free Hemoglobin value for verification
  • Ready-to-use format simplifies workflows
  • Recommended for monitoring quality performance
  • Packaged as 3 x 3 mL vials
  • Stored refrigerated at 2-8 °C
  • Critical for diagnosing hemolytic transfusion reactions and intravascular hemolysis
Catalog No. NC0998352 Supplier Catachem Supplier No. C46421
The CATACHEM HemoglobinCNTRL 1 is designed for monitoring the quality performance of plasma free hemoglobin determination procedures. This control material is essential for ensuring the accuracy and reliability of assays that measure free hemoglobin in plasma, particularly for diagnosing conditions like hemolytic transfusion reactions and intravascular hemolysis. Its ready-to-use liquid format simplifies laboratory workflows, eliminating the need for preparation. Laboratories are advised to use this control to maintain consistent quality performance in their analytical procedures.
  • Provides a known Plasma Free Hemoglobin value for verification
  • Ready-to-use format simplifies workflows
  • Recommended for monitoring quality performance
  • Packaged as 3 x 3 mL vials
  • Stored refrigerated at 2-8 °C
  • Critical for diagnosing hemolytic transfusion reactions and intravascular hemolysis