Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Catacxium Pd G4

Catalog No. 900349250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-034-9250MG 1 g
90-034-91G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-034-9250MG Supplier Sigma Aldrich Supplier No. 900349250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A fourth generation (G4) Buchwald precatalyst that is similar to the third generation (G3) precatalysts except that the amino group on the biphenyl backbone is methylated. This modification helps to prevent the limitations found when using the third generation precatalysts. It is air, moisture, and thermally-stable and shows good solubility in common organic solvents.

Specifications

CAS 68109-03-5
Molecular Formula C38H54NO3PPdS
Linear Formula C38H54NO3PPdS
Quantity 1 g
Molecular Weight (g/mol) 742.3 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.