Learn More
Strem, An Ascensus Company CAS# 1053658-91-5. 500MG. N-(2-Methoxyphenyl)-2-(di-t-butylphosphino)pyrrole, min. 95% [cataCXium POMetB]. MFCD06798301
SDP
Supplier:  Strem, An Ascensus Company 152975500MG
Encompass
CAS# 1053658-91-5. 500MG. N-(2-Methoxyphenyl)-2-(di-t-butylphosphino)pyrrole, min. 95% [cataCXium POMetB]. MFCD06798301. Molecular Formula: C19H28NOP. Molecular Weight: 317.41. Color/Form: white to yellowish pwdr. Stability: air sensitive. Strem# 15-2975. www.strem.com/catalog/v3/15-2975/
Catalog No. 50-196-3547