Learn More
Medchemexpress LLC HY-135849 100mg Medchemexpress, Catalase CAS:9001-05-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135849100MG
Encompass_Preferred
Medchemexpress, HY-135849 100mg Catalase CAS:9001-05-2 Catalase is a key enzyme in the metabolism of H2O2 and reactive oxygen species (ROS), and its expression and localization is markedly altered in tumors. Free oxygen radical scavenger. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1408