Learn More
Worthington Biochemical Corporation CATALASE 10X10
SDP
Catalog No. NC9840756
Encompass
Supplied as an aqueous solution of 2X crystallized catalase (Code: CTR without thymol) filtered through a 0.22 micron pore size membrane. Minimum of 30,000 units/ml; 10 ml/vial.
Catalog No. NC9840756 Supplier Worthington Biochemical Corporation Supplier No. LS001898
Supplied as an aqueous solution of 2X crystallized catalase (Code: CTR without thymol) filtered through a 0.22 micron pore size membrane. Minimum of 30,000 units/ml; 10 ml/vial.