Learn More
Key Scientific Products DROP-IT CATALASE 3%, 5 per Pack
SDP
Catalog No. NC0928259
Encompass_Preferred
A Catalase test is useful for the identification of many different bacteria. Since Streptococci lack the enzyme to catalyze hydrogen peroxide, and all Staphylococci possess it, 3% Catalase is used to separate the species. Also, Listeria monocytogenes, which is catalase positive can be differentiated from beta-hemolytic Streptococci. Most Neisseria species are catalase positive. 15% Catalase is used for anaerobes, with most anaerobes being catalase negative with the exceptions of several species in the B. fragilis group and a few of the anaerobic cocci. 30% Catalase is used with acid fast bacilli.
Catalog No. NC0928259 Supplier Key Scientific Products Supplier No. K980102
A Catalase test is useful for the identification of many different bacteria. Since Streptococci lack the enzyme to catalyze hydrogen peroxide, and all Staphylococci possess it, 3% Catalase is used to separate the species. Also, Listeria monocytogenes, which is catalase positive can be differentiated from beta-hemolytic Streptococci. Most Neisseria species are catalase positive. 15% Catalase is used for anaerobes, with most anaerobes being catalase negative with the exceptions of several species in the B. fragilis group and a few of the anaerobic cocci. 30% Catalase is used with acid fast bacilli.