Learn More
Selleck Chemical LLC Catalase Aspergillus niger E7315-100MG
SDP
Catalog No. 5000041843
Encompass_Preferred
Catalase Aspergillus niger is a key enzyme in the metabolism of H2O2 and reactive oxygen species (ROS) and its expression and localization is markedly altered in tumors Free oxygen radical scavenger
Catalog No. 50-000-41843 Supplier Selleck Chemical LLC Supplier No. E7315100MG
Catalase Aspergillus niger is a key enzyme in the metabolism of H2O2 and reactive oxygen species (ROS) and its expression and localization is markedly altered in tumors Free oxygen radical scavenger