Learn More
TRIBIOSCIENCE INC Catalase Assay 100 Tests
SDP
Catalog No. NC3942419
Encompass_Preferred
Tribioscience catalase Assay Kit provides a rapid, simple, reproducible, and sensitive tool for catalase activity determination in plasma, serum, urine, and other samples. The catalase activity assay uses enzyme reactions to produce color and fluorescence with the probe. The color intensity of the reaction at 570 nm or fluorescence intensity at λex/em = 530/590 nm is directly proportional to catalase concentration in the sample. The fluorometric assay is more sensitive than colorimetric assay. APPLICATIONS Direct Assays - Catalase enzyme activity in serum, plasma, urine, and other bio-samples. KEY FEATURES Flexible - Suitable for colorimetric and fluorometric methods. Accurate - Use 25 µL samples, detection ranges are 1.95-31.25 U/mL in 96-well plate the assay. Simple and high-throughput - Two-steps procedure - just load-incubate-adding-incubate-read. Kits can be used as a robust method.
Catalog No. NC3942419 Supplier TRIBIOSCIENCE INC Supplier No. TBS2006
Tribioscience catalase Assay Kit provides a rapid, simple, reproducible, and sensitive tool for catalase activity determination in plasma, serum, urine, and other samples. The catalase activity assay uses enzyme reactions to produce color and fluorescence with the probe. The color intensity of the reaction at 570 nm or fluorescence intensity at λex/em = 530/590 nm is directly proportional to catalase concentration in the sample. The fluorometric assay is more sensitive than colorimetric assay. APPLICATIONS Direct Assays - Catalase enzyme activity in serum, plasma, urine, and other bio-samples. KEY FEATURES Flexible - Suitable for colorimetric and fluorometric methods. Accurate - Use 25 µL samples, detection ranges are 1.95-31.25 U/mL in 96-well plate the assay. Simple and high-throughput - Two-steps procedure - just load-incubate-adding-incubate-read. Kits can be used as a robust method.