Learn More
MOLECULAR DEPOT LLC Catalase Bovine Liver -100 mg
Catalog No. 5000013360
Encompass_Preferred
Catalase from Bovine Liver (High Specific Activity) consists of native high specific activity (over 10 000 units/mg protein) Catalase purified from Bovine Liver through a series of chromatography steps For in vitro research use only
Catalog No. 50-000-13360 Supplier MOLECULAR DEPOT LLC Supplier No. B2010410
Catalase from Bovine Liver (High Specific Activity) consists of native high specific activity (over 10 000 units/mg protein) Catalase purified from Bovine Liver through a series of chromatography steps For in vitro research use only