Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Catalase Colorimetric Activity Kit

Catalog No. EIACATC
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIACATC 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIACATC Supplier Invitrogen™ Supplier No. EIACATC
Only null left
Add to Cart
Add to Cart

Includes

96-well plates, standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Assay Catalase Activity in Serum, Plasma, Cells, Tissues and Erythrocyte Lysates

The Catalase Activity research-use-only kit is a colorimetric activity assay designed for the quantification and detection of catalase activity in serum, plasma, cells, tissues and erythrocyte lysates.

This complete, ready-to-use kit includes clear 96 well plate(s), catalase standard (100 Unit/mL), catalase substrate, and other components to perform the assay. A 96-well microplate reader capable of reading optical density at 560 nm (acceptable range: 540-580 nm) is required for use of this kit.

Performance characteristics
• Assay type: colorimetric activity kit
• Sample types: serum, plasma, cells, tissues, and erythrocyte lysates.
• Sensitivity: 0.052 U/mL
• Standard curve range: 0.15–5.0 U/mL
• Reactivity: species independent

Background
Hydrogen peroxide is one of the most frequently occurring reactive oxygen species. It is formed either in the environment or as a by-product of aerobic metabolism, superoxide formation and dismutation, or as a product of oxidase activity. Both excessive hydrogen peroxide and its decomposition product hydroxyl radical, formed in a Fenton-type reaction, are harmful for most cell components. Its rapid removal is essential for all aerobically living prokaryotic and eukaryotic cells. However, hydrogen peroxide can act as a second messenger in signal transduction pathways, in immune cell activation, inflammation processes, cell proliferation, and apoptosis.

One of the most efficient ways of removing peroxide is through the enzyme catalase, which is encoded by a single gene and is highly conserved among species. Mammals, including humans and mice, express catalase in all tissues, and a high concentration of catalase can be found in the liver, kidneys, and erythrocytes. The expression is regulated at transcription, post-transcription, and post-translation levels. High catalase activity is detected in peroxisomes. More recently, short wavelength UV radiation has been shown to produce reactive oxygen species (ROS) through the action of catalase. This response is thought to act as a mechanism to protect DNA by converting damaging UV radiation into ROS species that can be metabolized and detoxified by cellular antioxidant enzymes. This assay has been validated for serum, EDTA and heparin plasmas from a variety of species.

Assay principle
The Catalase Activity kit is designed to quantitatively measure catalase activity in a variety of samples. A bovine catalase standard is provided to generate a standard curve for the assay and all samples should be read off of the standard curve. Samples are diluted in the provided Assay Buffer and added to the wells of a half area clear plate. Hydrogen peroxide is added to each well and the plate incubated at room temperature for 30 minutes. The supplied Colorimetric Detection Reagent is added, followed by diluted horseradish peroxidase, and incubated at room temperature for 15 minutes. The HRP reacts with the substrate in the presence of hydrogen peroxide to convert the colorless substrate into a pink-colored product. The colored product is read at 560 nm. Increasing levels of catalase in the samples causes a decrease in hydrogen peroxide concentration and a reduction in pink product.

Catalase unit definition
One unit of Catalase decomposes one micromole of hydrogen peroxide per minute at 25°C and pH 7.0.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 96-well plate(s), standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Store at 2–8°C.
Detection Method Colorimetric
Immunoassay Kit Format Assay Activity Kit
Product Type Catalase Colorimetric Activity Kit
Sample Type Plasma, Serum
For Use With (Equipment) Microplate Reader
Quantity 2 x 96 Tests
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Kit Named Catalase
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.