Learn More
Electron Microscopy Sciences Catalase Crystals
SDP
Catalog No. 5024925
Encompass_Preferred
Catalase crystals are mounted on a grid (3.05 mm) and negatively stained. They display very clear lattice plane spacings of approx. 8.75nm and 6.85nm. Ideal for high magnification calibration.
Catalog No. 50-249-25 Supplier Electron Microscopy Sciences Supplier No. 80014
Add to Cart
Add to Cart
Catalase crystals are mounted on a grid (3.05 mm) and negatively stained. They display very clear lattice plane spacings of approx. 8.75nm and 6.85nm. Ideal for high magnification calibration.