Learn More
Crescent Chemical Co Inc CATALASE FROM ASPERGILLUS
SDP
Numéro de catalogue. 50247776
Encompass_Preferred
Catalase from Aspergillus niger ca. 1800 U/mg lyophil. salt-free Catalase is used for the removal of peroxides, the generation of oxygen and, in coupled systems, for the determination of metabolites e.g. uric acid (1).A very stable preparation particularly suitable for immobilization. Siz
Numéro de catalogue. 50-247-776 Fournisseur Crescent Chemical Co Inc Code fournisseur SE26905.01
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Catalase from Aspergillus niger ca. 1800 U/mg lyophil. salt-free Catalase is used for the removal of peroxides, the generation of oxygen and, in coupled systems, for the determination of metabolites e.g. uric acid (1).A very stable preparation particularly suitable for immobilization. Siz
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.