Learn More
Crescent Chemical Co Inc CATALASE FROM BOVINE LIVER
SDP
Catalog No. 50247778
Encompass_Preferred
Catalase from bovine liver ca. 11 000 U/mg lyophil. salt-free Homogeneous in SDS-PAGE. Ca. 40 000 U/mg protein. Size - 1G Storage Conditions - -15 °C TO -25 °C Catalog Number - 26910.02 CAS 2593710
Catalog No. 50-247-778 Supplier Crescent Chemical Co Inc Supplier No. SE26910.02
Catalase from bovine liver ca. 11 000 U/mg lyophil. salt-free Homogeneous in SDS-PAGE. Ca. 40 000 U/mg protein. Size - 1G Storage Conditions - -15 °C TO -25 °C Catalog Number - 26910.02 CAS 2593710