Learn More
Athens Research And Technology Inc CATALASE HU ERYTHROCYTE 1MG
Catalog No. 50489683
Encompass_Preferred
Catalase, Human Erythrocyte: 1 MG; Frozen in 50 mM Tris-HCl, pH 8.0; MW 244,000; Extinction Coefficient=1.39; Purity: > =95% by SDS-PAGE; Activity: > 50,000 u/mg protein. pH 7.0. Prepared from whole blood in the Athens Research & Tech lab.
Catalog No. 50-489-683 Supplier Athens Research And Technology Inc Supplier No. 16050300001MG
Catalase, Human Erythrocyte: 1 MG; Frozen in 50 mM Tris-HCl, pH 8.0; MW 244,000; Extinction Coefficient=1.39; Purity: > =95% by SDS-PAGE; Activity: > 50,000 u/mg protein. pH 7.0. Prepared from whole blood in the Athens Research & Tech lab.