Learn More
Biomatik Corporation PLP1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07371
Encompass_Preferred
Plp1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PLP1. The Plp1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Myelin Proteolipid Protein (143-197Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PLP1. Accession Number: P60201. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6957