Learn More
Biomatik Corporation RAB3B anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506959
Encompass_Preferred
Rab3B Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RAB3B. The Rab3B Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ras-Related Protein Rab-3B Protein (1-219Aa). Applications: ELISA, WB, IHC. Target Synonyms: RAB3B. Accession Number: P20337. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6959 Supplier Biomatik Corporation Supplier No. CAC07373
Rab3B Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RAB3B. The Rab3B Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ras-Related Protein Rab-3B Protein (1-219Aa). Applications: ELISA, WB, IHC. Target Synonyms: RAB3B. Accession Number: P20337. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.