Learn More
Biomatik Corporation ROR1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07375
Encompass_Preferred
Ror1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ROR1. The Ror1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Inactive Tyrosine-Protein Kinase Transmembrane Receptor Ror1 Protein (30-391Aa). Applications: ELISA, IHC. Target Synonyms: ROR1. Accession Number: Q01973. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6961