Learn More
Biomatik Corporation METRNL anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07395
Encompass_Preferred
Metrnl Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to METRNL. The Metrnl Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Meteorin-Like Protein (46-311Aa). Applications: ELISA, WB. Target Synonyms: METRNL. Accession Number: Q641Q3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6981