Learn More
Biomatik Corporation NOSTRIN anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07397
Encompass_Preferred
Nostrin Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NOSTRIN. The Nostrin Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nostrin Protein (1-322Aa). Applications: ELISA, WB, IHC. Target Synonyms: NOSTRIN. Accession Number: Q8IVI9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6983