Learn More
Biomatik Corporation Afp anti-Mouse, Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07407
Encompass_Preferred
The Afp Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Human, Rat. Immunogen: Recombinant mouse Alpha-fetoprotein (19-605AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: Alpha-1-fetoprotein, Alpha-fetoglobulin, Afp. UniProt: P02772. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6993