Learn More
Biomatik Corporation APEX1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506994
Encompass_Preferred
The APEX1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human DNA- (apurinic or apyrimidinic site) lyase Protein (32-318AA). Applications: ELISA, WB, IHC, ChIP. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: APEX nuclease, Apurinic-apyrimidinic endonuclease 1, REF-1, Redox factor-1, APEX1, APE, APE1, APEX, APX, HAP1, REF1. UniProt: P27695. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6994 Supplier Biomatik Corporation Supplier No. CAC07408
The APEX1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human DNA- (apurinic or apyrimidinic site) lyase Protein (32-318AA). Applications: ELISA, WB, IHC, ChIP. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: APEX nuclease, Apurinic-apyrimidinic endonuclease 1, REF-1, Redox factor-1, APEX1, APE, APE1, APEX, APX, HAP1, REF1. UniProt: P27695. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.