Learn More
Biomatik Corporation APOA4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07409
Encompass_Preferred
The APOA4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Apolipoprotein A-IV protein (21-396AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: Apolipoprotein A4, ApoA-IV, Apolipoprotein A4, APOA4. UniProt: P06727. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6995