Learn More
Biomatik Corporation ATF3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506997
Encompass_Preferred
The ATF3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cyclic AMP-dependent transcription factor ATF-3 protein (1-171AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Cyclic AMP-dependent transcription factor ATF-3, cAMP-dependent transcription factor ATF-3, Activating transcription factor 3, ATF3. UniProt: P18847. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6997 Supplier Biomatik Corporation Supplier No. CAC07411
The ATF3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cyclic AMP-dependent transcription factor ATF-3 protein (1-171AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Cyclic AMP-dependent transcription factor ATF-3, cAMP-dependent transcription factor ATF-3, Activating transcription factor 3, ATF3. UniProt: P18847. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.