Learn More
Biomatik Corporation ATP6AP2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506998
Encompass_Preferred
The ATP6AP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Renin receptor protein (1-176AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: ATPase H (+)-transporting lysosomal accessory protein 2, ATPase H (+)-transporting lysosomal-interacting protein 2, ER-localized type I transmembrane adaptor, Embryonic liver differentiation factor 10, N14F, Renin/prorenin receptor, Vacuolar ATP synthase membrane sector-associated protein M8-9, HT028, MSTP009, PSEC0072, ATP6IP2, CAPER, ELDF10, ATP6AP2. UniProt: O75787. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6998 Supplier Biomatik Corporation Supplier No. CAC07412
The ATP6AP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Renin receptor protein (1-176AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: ATPase H (+)-transporting lysosomal accessory protein 2, ATPase H (+)-transporting lysosomal-interacting protein 2, ER-localized type I transmembrane adaptor, Embryonic liver differentiation factor 10, N14F, Renin/prorenin receptor, Vacuolar ATP synthase membrane sector-associated protein M8-9, HT028, MSTP009, PSEC0072, ATP6IP2, CAPER, ELDF10, ATP6AP2. UniProt: O75787. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.