Learn More
Biomatik Corporation TF anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07413
Encompass_Preferred
Tf Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TF. The Tf Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Human Transferrin (Native Protein). Applications: ELISA, WB. Target Synonyms: TF. Accession Number: P02787. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6999