Learn More
Biomatik Corporation GPI anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07415
Encompass_Preferred
The GPI Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Glucose-6-phosphate isomerase (2-554AA). Applications: ELISA, WB, IHC, IF, IP. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200, IP:1:200-1:2000. Alternative Names: Autocrine motility factor, Phosphoglucose isomerase, Phosphohexose isomerase, Sperm antigen 36. UniProt: P06744. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7001