Learn More
Biomatik Corporation EIF4A2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07416
Encompass_Preferred
The EIF4A2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Eukaryotic initiation factor 4A-II protein (1-407AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: EIF4A2, DDX2B, EIF4F, eIF-4A-II eIF4A-II, ATP-dependent RNA helicase eIF4A-2. UniProt: Q14240. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7002