Learn More
Biomatik Corporation CNTFR anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07419
Encompass_Preferred
The CNTFR Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Ciliary neurotrophic factor receptor subunit alpha protein (106-342AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:200-1000, IHC:1:20-1:200. Alternative Names: Ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR-alpha, CNTFR. UniProt: P26992. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7005