Learn More
Biomatik Corporation ELAVL2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07421
Encompass_Preferred
The ELAVL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human ELAV-like protein 2 protein (4-356AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: ELAV-like neuronal protein 1, Hu-antigen B, Nervous system-specific RNA-binding protein Hel-N1, ELAVL2, HUB. UniProt: Q12926. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7007