Learn More
Biomatik Corporation GABRA4 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07423
Encompass_Preferred
The GABRA4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Gamma-aminobutyric acid receptor subunit alpha-4 protein (36-258AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:200-1000, IHC:1:20-1:200. Alternative Names: Gamma-aminobutyric acid receptor subunit alpha-4, GABA (A) receptor subunit alpha-4, GABRA4. UniProt: P48169. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7009