Learn More
Biomatik Corporation IFIT5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07428
Encompass_Preferred
Ifit5 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IFIT5. The Ifit5 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon-Induced Protein With Tetratricopeptide Repeats 5 Protein (107-320Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: IFIT5. Accession Number: Q13325. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7014