Learn More
Biomatik Corporation TSPAN6 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507029
Encompass_Preferred
Tspan6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TSPAN6. The Tspan6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tetraspanin-6 Protein (115-208Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TSPAN6. Accession Number: O43657. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7029 Supplier Biomatik Corporation Supplier No. CAC07443
Tspan6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TSPAN6. The Tspan6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tetraspanin-6 Protein (115-208Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TSPAN6. Accession Number: O43657. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.