Learn More
Biomatik Corporation CTSB anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07449
Encompass_Preferred
The CTSB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Cathepsin B protein (82-333AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. UniProt: P07858. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7035