Learn More
Biomatik Corporation Cxcl5 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07452
Encompass_Preferred
The Cxcl5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Human. Immunogen: Recombinant mouse C-X-C motif chemokine 5 protein (46-117AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: Cytokine LIX Small-inducible cytokine B5. UniProt: P50228. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7038