Learn More
Biomatik Corporation NEU3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07457
Encompass_Preferred
Neu3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NEU3. The Neu3 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sialidase-3 Protein (2-428Aa). Applications: ELISA, WB. Target Synonyms: NEU3. Accession Number: Q9UQ49. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7043